missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen tetrachloroaurate(III) trihydrate, ACS, 99.99% (metals basis), Au 49.0% min

Catalog No. p-4407505
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3640003 1 g
AA3640006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3640003 Supplier Thermo Scientific Chemicals Supplier No. 03640003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16961-25-4 | AuCl4H | 339.77 g/mol

It is used for microanalysis of rubidium, cesium and in the determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in the control rods of nuclear reactors and in the manufacturing of electric light filaments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16961-25-4
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
MDL Number MFCD00149904
InChI Key VDLSFRRYNGEBEJ-UHFFFAOYSA-K
Synonym Chloroauric acid
IUPAC Name trichlorogold hydrochloride
SMILES Cl.Cl[Au](Cl)Cl

Specifications

Physical Form Crystalline
Quantity 1 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water. Insoluble in ether.
Formula Weight 393.83 (339.79 Anhydrous)
Grade ACS Reagent
Sensitivity Hygroscopic; Light sensitive
Chemical Name or Material Hydrogen tetrachloroaurate trihydrate, For analysis ACS
Hazard Category H302-H314-H317-H335
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 240-948-4
RTECSNumber MD5428000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only