missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min

Catalog No. AA1232503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1232503 1 g
AA1232506 5 g
AA1232514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1232503 Supplier Thermo Scientific Chemicals Supplier No. 01232503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for microanalysis of rubidium, cesium and determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in control rods in nuclear reactors and in the manufacture of light electric light filaments. | CAS: 27988-77-8 | AuCl4H | 339.77 g/mol

Hydrogen tetrachloroaurate(III) hydrate is used for microanalysis of rubidium, cesium and determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in control rods in nuclear reactors and in the manufacture of light electric light filaments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27988-77-8
MDL Number MFCD00149903
IUPAC Name hydrogen tetrachlorogoldtris(ylium)
SMILES [H+].Cl[Au+3](Cl)(Cl)Cl

Specifications

Packaging Glass bottle
Molecular Formula AuCl4H
Quantity 1 g
UN Number UN3260
Sensitivity Hygroscopic; Light sensitive
Synonym Tetrachloroauric(III) acid,Chloroauric acid
Solubility Information Soluble in water. Insoluble in ether.
InChI Key RFWCEGCWRSNAGT-UHFFFAOYSA-K
Molecular Weight (g/mol) 339.77
Formula Weight 339.79 (anhydrous)
Percent Purity 99.9%
Physical Form Crystalline
Chemical Name or Material Hydrogen tetrachloroaurate(III) hydrate
Show More Show Less
Hazard Category H302-H314-H317-H335
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 240-948-4
RTECSNumber MD5428000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only