missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen tetrachloroaurate hydrate, 99.999%, (trace metal basis)

Catalog No. AC437170010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437170010 1 g Glass bottle
AC437170050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437170010 Supplier Thermo Scientific Chemicals Supplier No. 437170010

CAS: 27988-77-8 | AuCl4H | 339.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis

Chemical Identifiers

CAS 27988-77-8
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
MDL Number MFCD00149903
InChI Key RFWCEGCWRSNAGT-UHFFFAOYSA-K
Synonym chloroauric acid hydrate, Gold trichloride acid hydrate
IUPAC Name hydrogen tetrachlorogoldtris(ylium)
SMILES [H+].Cl[Au+3](Cl)(Cl)Cl

Specifications

Melting Point 30.0°C
Color Yellow to Orange
Physical Form Crystals
Quantity 1 g
Assay Percent Range 49 to 51% (Au)
Linear Formula HAuCl4·xH2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 339.78
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material hydrogen tetrachloroaurate hydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

RUO – Research Use Only