missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide, Topical Solution, Contains 0.05% max. Acetophenetidin as a Preservative, 2.5 to 3.5%, U.S.P., J.T. Baker™

Catalog No. p-5391208
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Catalog No. Quantity Packaging
02003086 500 mL Narrow Mouth Amber Glass Bottle
02003078 4 L Narrow Mouth Amber Glass Bottle
2 options

Catalog No. 02003086

Supplier: Avantor AVR218201

Only null left

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Quantity 500 mL
Formula Weight 34.01
Percent Purity 2.5 to 3.5%
Grade USP
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.00kg
Chemical Name or Material Hydrogen Peroxide, Topical Solution, Contains 0.05% max. Acetophenetidin as a Preservative