missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide Solution, 29.0-32.0%, ACS, MilliporeSigma™

Catalog No. p-4382203
Change view
Click to view available options
Quantity:
500 mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MHX06353 500 mL
MHX06356 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MHX06353 Supplier Technidata Supplier No. HX06353

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
Assay Percent Range 29.0 to 32.0 %
Quantity 500 mL
Formula Weight 34.01
Physical Form Liquid
Chemical Name or Material Hydrogen Peroxide Solution, For ACS analysis