missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide Solution, 25-35%, ≥30% (RT), MilliporeSigma™ Supelco™

Catalog No. p-7187792
Change view
Click to view available options
Quantity:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018036 1 L
111017944 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111018036 Supplier Merck Emd Millipore Supplier No. 169111LF

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00011333

Specifications

Density 1.11 g/mL (at 20°C)
CAS 7722-84-1
Linear Formula H2O2
Quantity 1 L
Formula Weight 34.01
Vapor Pressure 23.3 mmHg (30°C)
Percent Purity ≥30% (RT)
Grade For ultratrace analysis
Concentration or Composition (by Analyte or Components) 25-35%
ShelfLife 5 years