missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide 30% LM Prime, Transene

Catalog No. 502744719
Encompass
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502744719 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 502744719 Supplier Doe and Ingalls Management LLC Supplier No. T0708168GAL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydrogen Peroxide 30% Technical is a powerful oxidizing agent widely used in industrial applications.

Hydrogen peroxide is a versatile product with many applications and uses and is a great solution for disinfecting surfaces and textiles. Hydrogen peroxide breaks down into oxygen and water leaving no toxic residues.

Chemical Identifiers

CAS 7722-84-1
Molecular Weight (g/mol) 34 g/mol

Specifications

Quantity 1 gal.
Packaging HDPE
Boiling Point 150.2°C
Percent Purity 30%
Grade Electronic
Chemical Name or Material Hydrogen Peroxide