missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen hexachloroplatinate(IV) hydrate, 99.9%, (trace metal basis), 38 to 40% Pt

Catalog No. p-3734983
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC363590010 1 g Glass bottle
AC363590050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC363590010 Supplier Thermo Scientific Chemicals Supplier No. 363590010
Only null left

CAS: 26023-84-7 | Cl6H2Pt | 409.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 26023-84-7
Molecular Formula Cl6H2Pt
Molecular Weight (g/mol) 409.80
MDL Number MFCD00149909
InChI Key ZKOQTCCVSRSECD-UHFFFAOYSA-J
Synonym Hexachloroplatinic acid hexahydrate, Platinic chloride hexahydrate
IUPAC Name dihydrogen hexachloroplatinumtetrakis(ylium)
SMILES [H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 60.0°C
Physical Form Chunks and/or Powder
Quantity 1 g
Assay Percent Range 38 to 40% (Pt)
Linear Formula H2PtCl6·xH2O
Merck Index 14, 7526
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 409.82
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Hydrogen hexachloroplatinate(IV) hydrate, 38 to 40% Pt
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only