missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen fluoride pyridine complex, ca 70% HF, Thermo Scientific Chemicals

Catalog No. aal1711714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1711714 25 g
AAL1711722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1711714 Supplier Thermo Scientific Chemicals Supplier No. L1711714
Only null left

Hydrogen fluoride pyridine complex is used to prepare beta-fluoroamines via reaction with amino alcohols. As a fluorinating agent, is involved in the fluorination of 2,2-diary-1,3-dithiolanes. It is associated with nitrosonium tetrafluorborate to prepare gem-difluorides by reacting with ketoximes. It is also used to convert secondary and tertiary alcohols into their respective alkyl fluorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydrogen fluoride pyridine complex is used to prepare beta-fluoroamines via reaction with amino alcohols. As a fluorinating agent, is involved in the fluorination of 2,2-diary-1,3-dithiolanes. It is associated with nitrosonium tetrafluorborate to prepare gem-difluorides by reacting with ketoximes. It is also used to convert secondary and tertiary alcohols into their respective alkyl fluorides.

Notes
Moisture sensitive. Store in cool place. Incompatible with strong bases, alkali metals, strong oxidizing agents, metals and strong acids. It reacts violently with water.

Chemical Identifiers

CAS 62778-11-4
Molecular Formula C5H6FN
Molecular Weight (g/mol) 99.108
MDL Number MFCD00012436
InChI Key GRJJQCWNZGRKAU-UHFFFAOYSA-N
Synonym pyridine hydrofluoride, hydrogen fluoride-pyridine, olah's reagent, pyridinium fluoride, hydrogen fluoride pyridine complex, hydrogen fluoride pyridine, hf-pyridine, pyridine, hydrofluoride 1:1, pyridinium poly hydrogen fluoride, hydrogen fluoride/pyridine
PubChem CID 64774
IUPAC Name pyridine;hydrofluoride
SMILES C1=CC=NC=C1.F

Specifications

Density 1.1
Boiling Point 50°C (1 mmHg)
Quantity 25 g
UN Number UN1790
Beilstein 4750151
Sensitivity Moisture sensitive
Merck Index 14,6821
Assay ≈70% HF
Chemical Name or Material Hydrogen fluoride pyridine complex
Hazard Category H300+H310+H330-H314-H335
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: HYDROFLUORIC ACID
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only