missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen fluoride-pyridine, approx. 70% HF, approx. 30% Pyridine

Catalog No. p-143902
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Plastic bottle
Plastic Bottle in Tin Can
Catalog No. Quantity Packaging
AC180070100 10 g Plastic bottle
AC180070500 50 g Plastic Bottle in Tin Can
AC180071000 100 g Plastic Bottle in Tin Can
3 options

Catalog No. AC180070100

Supplier: Thermo Scientific Chemicals 180070100

Only null left

CAS: 32001-55-1 | C5H6FN | 99.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 32001-55-1
Molecular Formula C5H6FN
Molecular Weight (g/mol) 99.11
MDL Number MFCD00012436
InChI Key GRJJQCWNZGRKAU-UHFFFAOYSA-N
Synonym pyridine hydrofluoride, hydrogen fluoride-pyridine, olah's reagent, pyridinium fluoride, hydrogen fluoride pyridine complex, hydrogen fluoride pyridine, hf-pyridine, pyridine, hydrofluoride 1:1, pyridinium poly hydrogen fluoride, hydrogen fluoride/pyridine
PubChem CID 64774
IUPAC Name pyridine;hydrofluoride
SMILES C1=CC=NC=C1.F

Specifications

Color Yellow to Brown
Density 1.1000g/mL
Assay Percent Range 65-70%
Packaging Plastic bottle
Quantity 10 g
Fieser 16,286; 16,050; 05,538; 06,473; 09,399; 12,419
Specific Gravity 1.1
Formula Weight 99.11
Percent Purity 65-70%
Physical Form Liquid
Chemical Name or Material Hydrogen fluoride-pyridine, 65-70%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
EINECSNumber 250-889-6
TSCA TSCA

RUO – Research Use Only