missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen chloride, pure, 5 to 6N solution in 2-propanol

Catalog No. p-4509480
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC133700010 1 L Plastic bottle
AC133700025 2.5 L Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC133700010 Supplier Thermo Scientific Chemicals Supplier No. 133700010
Only null left

CAS: 7647-01-0 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Hydrochloric acid
Color Colorless to Yellow
CAS 7647-01-0, 67-63-0
Physical Form Liquid
Quantity 1 L
Molecular Formula ClH
Linear Formula HCl
MDL Number MFCD00011324 MFCD00792839
Flash Point 11°C
Fieser 04,450; 05,533; 06,283
Merck Index 15,4831
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Solubility Information Solubility in water: >1000g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol: >50g/L
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name chlorane
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.45
Grade Pure
Concentration or Composition (by Analyte or Components) 5N min.
Packaging Plastic bottle
Specific Gravity 0.909
Density 0.9090g/mL
Show More Show Less

RUO – Research Use Only