missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen chloride, pure, 2N solution in diethyl ether, AcroSeal™

CAS: 7647-01-0 | ClH | 36.46 g/mol

$121.78 - $393.57

Specifications

Chemical Name or Material Hydrochloric acid
Color Colorless
Name Note 2N solution in diethyl ether
CAS 7647-01-0,60-29-7
Concentration 1.90 to 2.20N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC368471000
View Documents
Thermo Scientific Chemicals
368471000
100 mL AcroSeal™ Glass bottle
Each for $121.78
Only null left
 
AC368478000
View Documents
Thermo Scientific Chemicals
368478000
800 mL AcroSeal™ Glass Bottle
Each for $393.57
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hydrochloric acid
2N solution in diethyl ether
1.90 to 2.20N
0.74
ClH
Liquid
04,450; 05,533; 06,283
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
VEXZGXHMUGYJMC-UHFFFAOYSA-N
36.46
CHEBI:17883
Pure
Colorless
7647-01-0,60-29-7
0.7400g/mL
−40°C
HCl
MFCD00011324 MFCD00792839
10, 4701
Solubility in water: soluble. Other solubilities: soluble in most organic solvents
Cl
313
36.45
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only