missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen chloride, 5 to 6N solution in 2-propanol, AcroSeal™

CAS: 7647-01-0 | ClH | 36.46 g/mol

Supplier:  Thermo Scientific Chemicals 428608000

 View more versions of this product

Catalog No. AC428608000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hydrochloric acid
5 to 6N solution in 2-propanol
5N min.
AcroSeal™ Glass Bottle
11°C
ClH
HCl
14, 4794
Solubility in water: >1000g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol: >50g/L
Cl
36.46
CHEBI:17883
Colorless to Yellow
7647-01-0,67-63-0
0.9090g/mL
0.909
800 mL
Liquid
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
36.45
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only