missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen chloride, 5 to 6N solution in 2-propanol, AcroSeal™

Catalog No. AC428601000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428601000 100 mL AcroSeal™ Glass Bottle
AC428608000 800 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428601000 Supplier Thermo Scientific Chemicals Supplier No. 428601000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7647-01-0 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Hydrochloric acid
Name Note 5 to 6N solution in 2-propanol
CAS 7647-01-0, 67-63-0
Color Colorless to Yellow
Concentration 5N min.
Density 0.9090g/mL
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.909
Flash Point 11°C
Molecular Formula ClH
Linear Formula HCl
MDL Number MFCD00011324 MFCD00792839
Quantity 100 mL
Physical Form Liquid
Merck Index 14, 4794
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Solubility Information Solubility in water: >1000g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol: >50g/L
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name chlorane
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.45
Show More Show Less

RUO – Research Use Only