missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen chloride, 1.25M solution in ethanol, AcroSeal™
CAS: 7647-01-0 | ClH | 36.46 g/mol
$116.26 - $116.26
Specifications
| Chemical Name or Material | Hydrochloric acid |
|---|---|
| Name Note | 1.25M solution in ethanol |
| CAS | 7647-01-0,64-17-5 |
| Density | 0.8250g/mL |
| Packaging | AcroSeal™ Glass Bottle |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC427011000
|
Thermo Scientific Chemicals
427011000 |
100 mL | AcroSeal™ Glass Bottle |
Each for $116.26
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Hydrochloric acid | |
| 7647-01-0,64-17-5 | |
| AcroSeal™ Glass Bottle | |
| 12°C | |
| HCl | |
| 14, 4794 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 36.46 | |
| CHEBI:17883 |
| 1.25M solution in ethanol | |
| 0.8250g/mL | |
| 0.825 | |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| Cl | |
| 313 | |
| 36.46 |
For Manufacturing and Laboratory Use. Not for beverage use