missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Bromide - Methanol Reagent (5-10%) [for Esterification], TCI America™

Catalog No. x004325ml
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X004325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. X004325ML Supplier TCI America Supplier No. X004325ML
Only null left

Chemical Identifiers

CAS 10035-10-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
IUPAC Name hydrogen bromide
SMILES Br

Specifications

Color Yellow
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 2924
Chemical Name or Material Hydrogen Bromide - Methanol Reagent (5-10%) [for Esterification]
RTECSNumber MW3850000
TSCA Yes
Recommended Storage Refrigerator