missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Bromide, 33% Solution in Acetic Acid, 31 -35%, Spectrum™ Chemical

Catalog No. 18610944
Change view
Click to view available options
Quantity:
500 mL
1 L
100 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610944 100 kg Amber Glass Bottle
18610942 500 mL Amber Glass Bottle
18610943 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18610944 Supplier Spectrum Chemical Mfg Cor Supplier No. H1141100KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Hydrogen Bromide, 33 Percent Solution in Acetic Acid
CAS 64-19-7, 10035-10-6
Physical Form Liquid
Quantity 100 kg
Percent Purity 31 to 35%
Grade Ungraded
Packaging Amber Glass Bottle
Appearance Light to Dark Yellow Liquid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.