missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Bromide, 33% Solution in Acetic Acid, 31 -35%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor H11411LTGL
Specifications
| 64-19-7,10035-10-6 | |
| 31 to 35% | |
| Amber Glass Bottle |
| 1 L | |
| Reagent | |
| Light to Dark Yellow Liquid |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.