missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Bromide (30% in Acetic Acid, ca. 5.1mol/L) [for Peptide research], TCI America™
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 500 g |
| UN Number | 2920 |
| Formula Weight | 80.91 |
| Chemical Name or Material | Hydrogen Bromide (30% in Acetic Acid, ca. 5.1mol/L) [for Peptide research] |
Safety and Handling
| RTECSNumber | MW3850000 |
| TSCA | Yes |