missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Bromide (30% in Acetic Acid, ca. 5.1mol/L) [for Peptide research], TCI America™

Catalog No. H018225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H018225G 25 g
H0182500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H018225G Supplier TCI America Supplier No. H018225G

Chemical Identifiers

CAS 10035-10-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
IUPAC Name hydrogen bromide
SMILES Br

Specifications

Physical Form Liquid
Quantity 25 g
UN Number 2920
Formula Weight 80.91
Chemical Name or Material Hydrogen Bromide (30% in Acetic Acid, ca. 5.1mol/L) [for Peptide research]
RTECSNumber MW3850000
TSCA Yes