missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, Ultratrace, 47-51%, Spectrum™ Chemical

Catalog No. p-7165653
Change view
Click to view available options
Quantity:
500 mL
2 L
Packaging:
PTFE Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610940 500 mL PTFE Bottle
18610939 2 L PTFE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18610940 Supplier Spectrum Chemical Mfg Cor Supplier No. H1120500MLTF
Only null left

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
IUPAC Name hydrogen fluoride
SMILES F

Specifications

CAS Max % 1
Packaging PTFE Bottle
Quantity 500 mL
Percent Purity 47 to 51%
Grade High Purity Solvent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.