missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid (TraceMetal™ Grade), Fisher Chemical™

Catalog No. p-20226
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
HDPE Bottle
LDPE Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A513500 500 mL LDPE Bottle
A5134 4 L HDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A513500 Supplier Seastar Chemicals Inc Supplier No. S010501TFIW03
Only null left

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID 14917
ChEBI CHEBI:29228
IUPAC Name hydrogen fluoride
SMILES F

Specifications

pH 2.0
Color Colorless
Melting Point -35°C
Boiling Point 105°C
Packaging LDPE Bottle
Certificate of Analysis Certificate of Lot Analysis included with each shipment.
Quantity 500 mL
Physical Form Liquid
UN Number 1790
Merck Index 15, 4827
Formula Weight 20.01
Grade TraceMetal
Chemical Name or Material Hydrofluoric Acid
Show More Show Less
DOTInformation DOT Class 8, : Corrosive