missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, TraceGrade, 47-51%, Spectrum™ Chemical

Catalog No. p-7165654
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610938 500 mL Poly Bottle
18610937 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18610938 Supplier Spectrum Chemical Mfg Cor Supplier No. H1124500MLPL
Only null left

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
IUPAC Name hydrogen fluoride
SMILES F

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 500 mL
Percent Purity 47 to 51%
Grade High Purity Solvent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.