missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥48%, Honeywell Fluka™

Catalog No. p-7071961
Change view
Click to view available options
Quantity:
500 mL
1 L
5 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000780 500 mL
6000779 1 L
6000781 5 L
60045806 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6000780 Supplier Honeywell Inc Supplier No. 30107500ML
Only null left

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥48%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID 14917
ChEBI CHEBI:29228
SMILES F

Specifications

Concentration ≥48%
Density 1.15g/mL at 25°C
Residue after Ignition ≤0.0005% (as SO4)
Vapor Pressure 25mmHg (20°C)
Quantity 500 mL
UN Number 1790
IUPAC Name fluorane
Formula Weight 20.01g/mol
Grade ACS/puriss. p.a.
Chemical Name or Material Hydrofluoric Acid, Reag. Ph. Eur. reag. ISO
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P233
Precautionary Statement H300 + H310 + H330-H314
EINECSNumber 231-634-8