missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, OmniTrace™, MilliporeSigma™

Catalog No. MHX06271
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MHX06271 500 mL Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MHX06271 Supplier Technidata Supplier No. HX06271

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For General Chemistry

Recommended Storage

Store at room temperature

Chemical Identifiers

Molecular Formula HF in water

Specifications

Melting Point -35°C
Boiling Point 106°C (1013hPa)
Packaging Plastic Bottle
Quantity 500 mL
Recommended Storage Store at room temperature