missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric acid, Environmental Grade Plus, 47-51%, Thermo Scientific Chemicals

$2602.05 - $2602.05

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID 14917
ChEBI CHEBI:29228
SMILES F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA38746AP
View Documents
Thermo Scientific Chemicals
038746AP
500 mL
Each for $2,602.05
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7664-39-3
20.01
KRHYYFGTRYWZRS-UHFFFAOYSA-N
14917
F
FH
MFCD00011346
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
CHEBI:29228

Specifications

Colorless
112°C
Acrid
FH
MFCD00011346
14,4790
KRHYYFGTRYWZRS-UHFFFAOYSA-N
fluorane
14917
20.01
Environmental Plus
7664-39-3
1.16 g/mL
500 mL
Liquid, Double Sub-Boiling Quartz Distillation
UN1790
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
F
20.01
CHEBI:29228
47 to 51%
Hydrofluoric acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H314
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.

P234-P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c

H290-H300+H310+H330-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROFLUORIC ACID

EINECSNumber : 231-634-8

RTECSNumber : MW7875000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only