missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric acid, ACS, 48-51%, Thermo Scientific™

Catalog No. AA33258LB2
Change view
Click to view available options
Quantity:
470mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33258LB2 470mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA33258LB2 Supplier Thermo Scientific Chemicals Supplier No. 33258LB2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID 14917
ChEBI CHEBI:29228
IUPAC Name hydrogen fluoride
SMILES [H+].[F-]

Specifications

Density 1.16
Odor Acrid
Sensitivity Hygroscopic
Quantity 470mL
UN Number UN1790
Merck Index 14,4790
Solubility Information Very soluble in waterMiscible with water and alcohol. Slightly miscible with ether.
Formula Weight 20.01
Percent Purity 48 to 51%
Grade ACS
Chemical Name or Material Hydrofluoric acid
Show More Show Less
Hazard Category H290-H300+H310+H330-H314-H335
Hazard Statement GHS H Statement
H300-H310-H330-H314; Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage.
Precautionary Statement P234-P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 231-634-8
RTECSNumber MW7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only