missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, ACS, 48-51%, Spectrum™ Chemical

H1045, 7664-39-3, HF

Supplier:  Spectrum Chemical Mfg Cor H1045500MLCS6

 View more versions of this product

Catalog No. 18603240

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Hydrofluoric Acid, 48 Percent, Suitable for Electronic Use, Reagent, ACS is a highly corrosive solution of hydrogen fluoride in water is capable of dissolving a wide range of materials including oxides and glass. Used in a variety of fields including oil refining and production of fluorides it can also be used to dissolve rock samples before analysis. As an ACS grade Reagent, Spectrum Chemical manufactured hydrofluoric acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Specifications

Specifications

7664-39-3
0.5 ppm
5 ppm
FH
UN1790
F
20.01
ACS
100%
Poly Bottle
6 x 500 mL
MFCD00011346
KRHYYFGTRYWZRS-UHFFFAOYSA-N
hydrogen fluoride
48 to 51%

Chemical Identifiers

7664-39-3
20.01
KRHYYFGTRYWZRS-UHFFFAOYSA-N
F
FH
MFCD00011346
hydrogen fluoride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions