missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid 5% LM PrimeHydrofluoric acid 5%, CMOS for the electronics industry, Transene

Catalog No. 502744714
Encompass
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502744714 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 502744714 Supplier Doe and Ingalls Management LLC Supplier No. T0605HFGAL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Primarily used for cleaning and etching silicon wafers in semiconductor manufacturing, removing oxide layers on metals, and other electronic cleaning applications.

Chemical Identifiers

CAS 7664-39-3
Molecular Weight (g/mol) 20.01 g/mol

Specifications

Packaging HDPE
Quantity 1 gal.
Percent Purity 5%
Grade Electronic
Chemical Name or Material Hydrofluoric Acid