missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid 49% LM Prime, Transene

Catalog No. p-200106339
Change view
Click to view available options
Quantity:
55 Gallon
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502744721 55 Gallon
502744722 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 502744721 Supplier Doe and Ingalls Management LLC Supplier No. T0708686GAL
Only null left

Hydrofluoric acid 49%, CMOS for the electronics industry

In the semiconductor field, it is mainly used for

  • Wafer surface cleaning
  • Cleaning and etching during chip processing
  • Other processes
  • In the field of new displays, it is mainly used for glass substrate cleaning, silicon nitride, silicon dioxide etching, etc
  • In the field of photovoltaic solar cells, it is mainly used for surface cleaning, etching edge removal, cleaning and velvet making of silicon wafers.

Chemical Identifiers

CAS 7664-39-3
Molecular Weight (g/mol) 20.01 g/mol

Specifications

Packaging HDPE
Quantity 55 Gallon
Percent Purity 49%
Grade Electronic
Chemical Name or Material Hydrofluoric Acid