missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, 49%, Electronic/Cleanroom Grade, 48.8-49.2%, Spectrum™ Chemical

Catalog No. 18610936
Change view
Click to view available options
Quantity:
1 gal.
55 gal.
Packaging:
Amber Glass Bottle
Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610936 55 gal. Drum
18610935 1 gal. Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18610936 Supplier Spectrum Chemical Mfg Cor Supplier No. H104355GLBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7664-39-3 , 7382-15-5
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
IUPAC Name hydrogen fluoride
SMILES F

Specifications

CAS Max % 46 to 52%, 48 to 54%
Packaging Drum
Quantity 55 gal.
Percent Purity 48.8 to 49.2%
Grade Electronic/Cleanroom
Chemical Name or Material Hydrofluoric Acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.