missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, 48-52%, Technical, Spectrum™ Chemical

H1053, 7664-39-3, HF

$530.43 - $6084.20

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
IUPAC Name hydrogen fluoride
SMILES F
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
18603244
View Documents
Spectrum Chemical Mfg Cor
H1053500ML
500 mL
Each for $530.43
Only null left
 
18603245
View Documents
Spectrum Chemical Mfg Cor
H1053500MLCS6
6 x 500 mL
Case of 6 Each for $2,355.83
Only null left
 
18603246
View Documents
Spectrum Chemical Mfg Cor
H10531GL
1 gal.
Each for $1,827.23
Only null left
 
18603247
View Documents
Spectrum Chemical Mfg Cor
H10531GLCS4
4 x 1 gal.
Case of 4 Each for $6,084.20
Only null left
 
Description

Description

Spectrum™ Chemical Hydrofluoric Acid, 48-52 Percent, Technical is a highly corrosive solution of hydrogen fluoride in water is capable of dissolving a wide range of materials including oxides and glass. Used in a variety of fields including oil refining and production of fluorides it can also be used to dissolve rock samples before analysis.

Specifications

Chemical Identifiers

7664-39-3
20.01
KRHYYFGTRYWZRS-UHFFFAOYSA-N
F
FH
MFCD00011346
hydrogen fluoride

Specifications

7664-39-3,7732-18-5
46 to 52%,48 to 54%
FH
UN1790
F
20.01
Technical
112°C
500 mL
MFCD00011346
KRHYYFGTRYWZRS-UHFFFAOYSA-N
hydrogen fluoride
48 to 52%
Hydrofluoric Acid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'