missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, 48-50% (Technical), Fisher Chemical

Catalog No. A14610LB
Change view
Click to view available options
Quantity:
10 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14610LB 10 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14610LB Supplier Thermo Fisher Scientific Supplier No. FLA14610LB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

pH 2.0
Color Colorless
Melting Point -35°C
CAS 7664-39-3, 7732-18-5
Boiling Point 105°C
Density 2.21 (Air = 1.0)
Identification Pass Test
Packaging Poly Bottle
Assay Percent Range 48 to 50 w/w %
Quantity 10 lb.
Molecular Formula FH
Physical Form Liquid
MDL Number MFCD00011346
UN Number 1790
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
SMILES F
IUPAC Name hydrogen fluoride
Molecular Weight (g/mol) 20.01
PubChem CID 14917
ChEBI CHEBI:29228
Formula Weight 20.01
Percent Purity 48 to 50% (w/w)
Grade Technical
Chemical Name or Material Hydrofluoric Acid, 48-50%
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
May be fatal if inhaled, absorbed through skin, or swallowed. Corrosive to metals. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:4
Flammability:0
Instability:1
DOTInformation DOT Class 8, : Corrosive