missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, 40.0%, Suprapur™, MilliporeSigma™

Catalog No. M1003352500
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1003352500 2.5 L
M1003350500 500 mL
M1003351000 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1003352500 Supplier Technidata Supplier No. 1003352500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym Hydrofluoric acid solution, Fluoric acid
IUPAC Name hydrogen fluoride
SMILES F

Specifications

Melting Point −44°C
Boiling Point 112°C (1013 hPa)
Density 1.13 g/cm3 (at 20°C)
Packaging Plastic Bottle
Assay Percent Range ≥ 40%
Quantity 2.5 L
Recommended Storage Store at 5 to 30°C