missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone Sodium Phosphate, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022877
Change view
Click to view available options
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111022877 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022877 Supplier Merck Emd Millipore Supplier No. BP188

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

CAS 6000-74-4
Quantity 100 mg
Molecular Formula C21H29Na2O8P
Linear Formula C21H29Na2O8P
Synonym Hydrocortisone 21-phosphate sodium salt; 11 beta,17 alpha,21-Trihydroxy-4-pregnene-3,20-dione 21-phosphate; 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione 21-phosphate; Cortisol 21-phosphate sodium salt
Formula Weight 486.40
Grade BP