missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014353
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014353 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014353 Supplier Merck Emd Millipore Supplier No. PHR1014500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Hydrocortisone is a degradation product of the corticosteroid, hydrocortisone acetate. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 50-23-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Linear Formula C21H30O5
MDL Number MFCD00011654
Melting Point 211°C to 214°C
Synonym 11 beta,17 alpha,21-Trihydroxypregn-4-ene-3,20-dione; 17-Hydroxycorticosterone; 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione; Cortisol; Kendall′s compound F; Reichstein′s substance M
Type Certified Reference Material
Quantity 500 mg
Formula Weight 362.46
Grade Certified Reference Material