missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone acetate, 97+%

CAS: 50-03-3 | C23H32O6 | 404.50 g/mol

Supplier:  Thermo Scientific Chemicals A1808903

 View more versions of this product

Catalog No. AAA1808903

Only null left
Explore available promotions

Description

Description

Hydrocortisone acetate is used to treat hemorrhoids and itching/swelling in the rectum and anus. It is used to treat certain intestinal problems (such as ulcerative colitis of the rectum and other rectal/anal inflammatory conditions).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store in the dark.
Specifications

Specifications

Hydrocortisone acetate
∼220°C (decomposition)
MFCD00037714
2066841
14,4787
Insoluble in water.
CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
404.50
CHEBI:17609
≥97%
50-03-3
C23H32O6
1 g
Light sensitive
hydrocortisone acetate, cortell, cortisol 21-acetate, cortisol acetate, cortacream, hydrocortisat, abbocort, bambicort, biocortar, chemysone
ALEXXDVDDISNDU-JZYPGELDSA-N
[2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
5744
404.51

Chemical Identifiers

50-03-3
404.50
ALEXXDVDDISNDU-JZYPGELDSA-N
5744
CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
C23H32O6
MFCD00037714
hydrocortisone acetate, cortell, cortisol 21-acetate, cortisol acetate, cortacream, hydrocortisat, abbocort, bambicort, biocortar, chemysone
CHEBI:17609
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

P201-P202-P260-P281-P308+P313-P501c

H360-H373

EINECSNumber : 200-004-4

RTECSNumber : GM8960000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only