missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone, 98%

Catalog No. aaa1629203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1629203 1 g
AAA1629206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1629203 Supplier Thermo Scientific Chemicals Supplier No. A1629203
Only null left

CAS: 50-23-7 | C21H30O5 | 362.466 g/mol

Cortisol, steroid hormone is released in response to stress and a low level of blood glucocorticoids. Its primary functions is to increase blood sugar through gluconeogenesis, suppress the immune system, and aid in fat, protein and carbohydrate metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cortisol, steroid hormone is released in response to stress and a low level of blood glucocorticoids. Its primary functions is to increase blood sugar through gluconeogenesis, suppress the immune system, and aid in fat, protein and carbohydrate metabolism.

Solubility
It is soluble in DMSO and ethanol.

Notes
Protect against light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 50-23-7
Molecular Formula C21H30O5
MDL Number MFCD00011654
InChI Key JYGXADMDTFJGBT-VWUMJDOOSA-N
Synonym hydrocortisone, cortisol, acticort, cetacort, cortef, 17-hydroxycorticosterone, hydrocortisyl, hydrasson, cobadex, cortril
PubChem CID 5754
ChEBI CHEBI:17650

Specifications

Melting Point 213°C to 215°C
Odor Odorless
Assay Percent Range 98%
Quantity 1 g
Beilstein 1354819
Merck Index 14,4787
Solubility Information It is soluble in DMSO and ethanol.
SMILES CC12CCC(=O)C=C1CCC3C2C(CC4(C3CCC4(C(=O)CO)O)C)O
IUPAC Name (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 362.466
Formula Weight 362.47
Percent Purity 98%
Chemical Name or Material Hydrocortisone
Show More Show Less
Hazard Category H360Df-H373
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
EINECSNumber 200-020-1
RTECSNumber GM8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only