missing translation for 'onlineSavingsMsg'
Learn More

Hydrocinnamic acid, 99%

Catalog No. AC159100050
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159100050 5 g Glass bottle
AC159101000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC159100050 Supplier Thermo Scientific Chemicals Supplier No. 159100050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 501-52-0 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 501-52-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00002771
InChI Key XMIIGOLPHOKFCH-UHFFFAOYSA-N
Synonym hydrocinnamic acid, 3-phenylpropionic acid, benzylacetic acid, benzenepropanoic acid, dihydrocinnamic acid, benzenepropionic acid, phenylpropanoic acid, 3-phenyl-propionic acid, phenylpropanoate, beta-phenylpropionic acid
PubChem CID 107
ChEBI CHEBI:28631
IUPAC Name 3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CCC(=O)O

Specifications

Melting Point 47.0°C to 50.0°C
Density 1.0700g/mL
Boiling Point 279.0°C to 281.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2CH2CO2H
Quantity 5 g
Beilstein 09, 508
Specific Gravity 1.07
Merck Index 15, 4822
Solubility Information Solubility in water: 0.5% in water; more soluble in hot water. Other solubilities: soluble in glacial acetic acid,soluble in alcohol,benzene,ether,chloroform,,petroleum ether and carbon disulfide
Formula Weight 150.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Hydrocinnamic acid, 99%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-924-5
RTECSNumber DA8600000
TSCA TSCA

RUO – Research Use Only