missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 6.00 N, Ricca Chemical
Supplier: Ricca Chemical Company 375055
Specifications
| Hydrochloric Acid | |
| 62.0,36.27 | |
| Poly Drum | |
| ClH | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| [H+].[Cl-] | |
| 36.46 | |
| CHEBI:17883 |
| 7647-01-0,7732-18-5 | |
| 64.0,37.75 | |
| 55 gal. | |
| MFCD00011324 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| 6.00N |