missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, 5% v/v aq. soln.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Yellow |
| Concentration | 5% v/v aq. soln. |
| Quantity | 1 L |
| Physical Form | Liquid |
| UN Number | UN1789 |
| IUPAC Name | palladium(2+) dichloride |
| Formula Weight | 36.46 |
| Chemical Name or Material | Hydrochloric acid |
Safety and Handling
| Hazard Category | H290-H315-H318 |
| Precautionary Statement | P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390 |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HYDROCHLORIC ACID |
| EINECSNumber | 231-595-7 |
| RTECSNumber | MW4025000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only