missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 1N standard solution

CAS: 7647-01-0 | ClH | 36.46 g/mol

$119.74 - $534.14

Specifications

Chemical Name or Material Hydrochloric acid
Color Colorless
Name Note 1N standard solution
CAS 7647-01-0,7732-18-5
Concentration 0.998 to 1.002N (20°C)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC124210010
View Documents
Thermo Scientific Chemicals
124210010
1 L Plastic Bottle
Each for $119.74
Only null left
 
AC124210025
View Documents
Thermo Scientific Chemicals
124210025
2.5 L Plastic Bottle
Each for $154.00
Only null left
 
AC124210100
View Documents
Thermo Scientific Chemicals
124210100
10 L Plastic drum
Each for $534.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
Specifications

Specifications

Hydrochloric acid
1N standard solution
0.998 to 1.002N (20°C)
ClH
Liquid
04,450; 05,533; 06,283
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
313
Colorless
7647-01-0,7732-18-5
1.0200g/mL
HCl
MFCD00011324 MFCD00792839
15, 4818
VEXZGXHMUGYJMC-UHFFFAOYSA-N
36.46
CHEBI:17883
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only