Learn More
Thermo Scientific™ Hydrocarbon Soluble Siliconizing Fluid
Catalog No. PI42800
Encompass
Change view
Click to view available options
Quantity:
120 mL
Catalog No. Quantity
PI42800 120 mL
1 options
Material is flammable before film is formed and HCl fumes are generated in its application. Fluid is acidic during application and care should be taken to avoid corrosion of metal that comes into contact with the liquid. After application, the surface is neutral.
Catalog No. PI42800 Supplier Thermo Scientific™ Supplier No. TS42800
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Material is flammable before film is formed and HCl fumes are generated in its application. Fluid is acidic during application and care should be taken to avoid corrosion of metal that comes into contact with the liquid. After application, the surface is neutral.
For siliconizing glass surfaces use Thermo Scientific™ Hydrocarbon Soluble Siliconizing Fluid.
For siliconizing glass surfaces use Thermo Scientific™ Hydrocarbon Soluble Siliconizing Fluid. This short chain, clear polymeric silicone fluid consists primarily of dichlorooctamethyltetrasiloxane and is soluble in organic solvents. When applied to glass, quartz or similar products, the unhydrolyzed chlorines present on the chain react with surface silanols to form a neutral, hydrophobic and tiightly bonded film over the entire surface. It is excellent for modifying metals, certain plastics, ceramics and fiber optics.
Additional Product Information
Hydrocarbon Soluble Siliconizing Fluid is acidic and care should be taken to avoid corrosion of metal that comes into contact with the liquid.
The fluid is acidic only during application; after application, the surface is neutral.
Specifications
For Use With GC
Hazards Corrosive to Eyes/Skin/Respiratory Tract
Standard Components 1,7-Dichloro-1,1,3,3,5,5,7,7-Octamethyltetrasiloxane
Quantity 120 mL
Product Type Hydrocarbon Soluble Siliconizing Fluid
For Research Use Only. Not for use in diagnostic procedures.