missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid, purum p.a., Honeywell™

Catalog No. p-7071840
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000696 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000696 Supplier Honeywell Chemicals Supplier No. 187301L
Only null left

Purum p.a.,≥48%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Chemical Identifiers

CAS 10035-10-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
IUPAC Name bromane
SMILES Br

Specifications

Density 1.49g/mL at 25°C
Vapor Pressure 8mmHg (25°C), 320psi (21.1°C)
Quantity 1 L
UN Number 1788
Beilstein 3587158
Formula Weight 80.91g/mol
Percent Purity ≥48%
Grade purum p.a.
Chemical Name or Material Hydrobromic Acid
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H314-H335
EINECSNumber 233-113-0
RTECSNumber MW3850000