missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid, ACS Reagent, Honeywell Fluka™

Catalog No. p-7071937
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000695 500 mL
6000693 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000695 Supplier Honeywell Chemicals Supplier No. 244260500ML
Only null left

ACS Reagent, 48%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10035-10-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
SMILES Br

Specifications

Concentration 48%
Density 1.49g/mL at 25°C
Residue after Ignition ≤0.002%
Vapor Pressure 8mmHg (25°C), 320psi (21.1°C)
Quantity 500 mL
UN Number 1788
Beilstein 3587158
IUPAC Name bromane
Formula Weight 80.91g/mol
Grade ACS Reagent
Chemical Name or Material Hydrobromic Acid
Show More Show Less
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H314-H335