missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic acid, 48% w/w aq. soln.

Catalog No. p-4408621
Change view
Click to view available options
Quantity:
4 x 940 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA14036AZ 4 x 940 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA14036AZ Supplier Thermo Scientific Chemicals Supplier No. 014036AZ
Only null left

CAS: 10035-10-6 | BrH | 80.91 g/mol

Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes and medicines. It is also used in the production of bromine compounds. It is used as an important agent in the industrial processes of isomerization, polymerization, hydration and dehydration and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10035-10-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
IUPAC Name hydrogen bromide
SMILES Br

Specifications

Density 1.49 g/mL
Sensitivity Air Sensitive
Quantity 4 x 940 mL
Physical Form Liquid
Beilstein 3587158
Solubility Information Miscible with water,alcohol and organic solvents.
Formula Weight 80.92
Chemical Name or Material Hydrobromic acid
Hazard Category H290-H314-H335
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 233-113-0
RTECSNumber MW3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only