missing translation for 'onlineSavingsMsg'
Learn More

Hydrindantin Hydrate, Approx. 96%, Spectrum™ Chemical

Catalog No. 18603161
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603161 5 g Amber Glass Bottle
18603162 25 g Amber Glass Bottle
18603163 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18603161 Supplier Spectrum Chemical Mfg Cor Supplier No. H24515GM
Get Up to $500 in Chemicals with Purchase
View all promotions

H2451, 5950-69-6, C18H10O6·2H2O

Spectrum™ Chemical Hydrindantin Hydrate

Chemical Identifiers

CAS 5950-69-6
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
InChI Key QHVADKNWNMILPQ-UHFFFAOYNA-N
IUPAC Name 2,2',3,3,3',3'-hexahydroxy-1H,1'H,2H,2'H,3H,3'H-[2,2'-biindene]-1,1'-dione
SMILES OC1(O)C2=CC=CC=C2C(=O)C1(O)C1(O)C(=O)C2=CC=CC=C2C1(O)O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity ∼96%
Grade Reagent