missing translation for 'onlineSavingsMsg'
Learn More

Hydrindantin dihydrate, 96%

Catalog No. AC159280100
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159280100 10 g Glass bottle
AC159280500 50 g Glass bottle
AC159282500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC159280100 Supplier Thermo Scientific Chemicals Supplier No. 159280100

CAS: 5950-69-6 | C18H14O8 | 358.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5950-69-6
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.29
MDL Number MFCD00149242
InChI Key QHVADKNWNMILPQ-HOTGVXAUSA-N
Synonym 2s-2,3,3-trihydroxy-2-2s-1,1,2-trihydroxy-3-oxoinden-2-yl inden-1-one
PubChem CID 6560392
IUPAC Name (2S)-2,3,3-trihydroxy-2-[(2S)-1,1,2-trihydroxy-3-oxoinden-2-yl]inden-1-one
SMILES C1=CC=C2C(=C1)C(=O)C(C2(O)O)(C3(C(=O)C4=CC=CC=C4C3(O)O)O)O

Specifications

Melting Point >240°C
Color Beige to Beige-Pink
Infrared Spectrum Authentic
Assay Percent Range 95% min
Packaging Glass bottle
Quantity 10 g
Merck Index 14, 4775
Solubility Information Solubility in water: insoluble
Formula Weight 358.29
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Hydrindantin dihydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 227-713-1

RUO – Research Use Only