missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrazine Sulfate, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
H1018, 10034-93-2, H4N2·H2SO4
Supplier: Spectrum Chemical Mfg Cor H1018125GM
Description
Spectrum™ Chemical Hydrazine Sulfate, Crystal, Reagent, ACS has been promoted to treat colon and rectal (colorectal) cancer, lung cancer, brain cancer (neuroblastoma), Hodgkin′s disease (lymph cancer), and other cancers. It is also used for the general weight loss and wasting (cachexia) associated with cancer. However, hydrazine sulfate is not approved for use with cancer patients in the United States. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Specifications
| 10034-93-2 | |
| Poly Bottle | |
| 125 g | |
| ZGCHATBSUIJLRL-UHFFFAOYSA-N | |
| hydrazine; sulfuric acid | |
| 99% |
| 100% | |
| H6N2O4S | |
| UN1759 | |
| NN.OS(O)(=O)=O | |
| 130.12 | |
| ACS |
Chemical Identifiers
| 10034-93-2 | |
| 130.12 | |
| hydrazine; sulfuric acid |
| H6N2O4S | |
| ZGCHATBSUIJLRL-UHFFFAOYSA-N | |
| NN.OS(O)(=O)=O |