Learn More
Hydrazine sulfate, 99+%
CAS: 10034-93-2 | H6N2O4S | 130.118 g/mol
Supplier: Thermo Scientific Chemicals A1079230
Description
Hydrazine sulfate is used in analytical chemistry for the gravimetric estimation of nickel, cobalt and cadmium. It is also used in the refining of rare metals as well as in the separation of polonium from tellurium. It finds application in the synthesis of chemical intermediate and organic compounds. Further, it acts as an antioxidant in soldering flux for light metals and as a precursor to hydrazine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHydrazine sulfate is used in analytical chemistry for the gravimetric estimation of nickel, cobalt and cadmium. It is also used in the refining of rare metals as well as in the separation of polonium from tellurium. It finds application in the synthesis of chemical intermediate and organic compounds. Further, it acts as an antioxidant in soldering flux for light metals and as a precursor to hydrazine.
Solubility
Soluble in water. Insoluble in alcohol.
Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.comIncompatible with oxidizing agents and bases.
Specifications
| Hydrazine sulfate | |
| 254°C | |
| 1.3 | |
| H6N2O4S | |
| 250 g | |
| 14,4772 | |
| Soluble in water. Insoluble in alcohol. | |
| NN.OS(=O)(=O)O | |
| 130.118 | |
| 130.12 |
| 10034-93-2 | |
| 1.37 | |
| Odorless | |
| MFCD00044873 | |
| UN3288 | |
| hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate | |
| ZGCHATBSUIJLRL-UHFFFAOYSA-N | |
| hydrazine;sulfuric acid | |
| 24842 | |
| ≥99% |
Chemical Identifiers
| 10034-93-2 | |
| 130.118 | |
| ZGCHATBSUIJLRL-UHFFFAOYSA-N | |
| 24842 | |
| NN.OS(=O)(=O)O |
| H6N2O4S | |
| MFCD00044873 | |
| hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate | |
| hydrazine;sulfuric acid |
Safety and Handling
GHS H Statement
H311-H331-H350-H302-H317
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Harmful if swallowed.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P333+P313-P361-P363-P501c
H301+H311+H331-H317-H350
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber : 233-110-4
RTECSNumber : MV9625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only